Learn More
Hexa-n-butylditin, 97%
Description
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 1.152 |
| Boiling Point | 197°C to 198°C (10 mmHg) |
| Flash Point | 124°C (255°F) |
| Linear Formula | (CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3 |
| Refractive Index | 1.512 |
| Quantity | 250 g |
| UN Number | UN2788 |
| Beilstein | 3605476 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Immiscible with water. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.