missing translation for 'onlineSavingsMsg'
Learn More

Hexa-n-butylditin, 97%

Product Code. 11411900
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411900 25 g 25g
11494054 10 g 10g
11404064 50 g 50g
11414064 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11411900 Supplier Thermo Scientific Alfa Aesar Supplier No. A12007.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 813-19-4 | C24H54Sn2 | 580.12 g/mol

Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-19-4
Molecular Formula C24H54Sn2
Molecular Weight (g/mol) 580.12
MDL Number MFCD00009417
InChI Key REDSKZBUUUQMSK-UHFFFAOYSA-N
Synonym hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
PubChem CID 6327815
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC

Specifications

Density 1.152
Boiling Point 197°C to 198°C (10 mmHg)
Flash Point 124°C
Linear Formula (CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
Refractive Index 1.512
Quantity 25 g
UN Number UN2788
Beilstein 3605476
Sensitivity Air and moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 580.07
Percent Purity 98%
Chemical Name or Material Hexa-n-butylditin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.