Learn More
Labconco Hex Rotor - CentriVap Pro
GREENER_CHOICE
Product Code. p-200132243 Shop All Labconco Corporation Products
Click to view available options
Unit Size:
Each
Material (Coating):
PTFE
Product Code. 30533446
Brand: Labconco 7555600
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Stainless steel and PTFE coated options available.
  • Compatible with CentriVap Pro Vacuum Concentrators
  • Stackable design
  • Stainless steel material\
  • Optional PTFE coated
Specifications
Material Stainless Steel
Type Rotor
Rotor System CentriVap Pro
Dimensions (L x W x H) 26.42 x 22.86 x 6.6 cm (10.4 x 9 x 2.6 in.)