missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hex Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529154
This item is not returnable.
View return policy
Description
Recommended positive controls: K562, THP-1. Predicted reactivity: Mouse (92%), Rat (94%), Pig (95%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.
Specifications
| Hex | |
| Polyclonal | |
| Unconjugated | |
| HHEX | |
| hematopoietically expressed homeobox; hematopoietically expressed homeobox, related sequence 2; hematopoietically-expressed homeobox protein Hhex; HEX; Hex1; HHEX; Hhex-rs2; HMPH; homeobox; homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; HOX11L-PEN; mHex; Prh; Prhx; proline-rich homeodomain-containing transcription factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3087 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q03014 | |
| HHEX | |
| Recombinant protein encompassing a sequence within the center region of human Hex. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction