missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heterogeneous Nuclear Ribonucleoprotein (A1-like) Antibody (S05-8G1), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | Heterogeneous Nuclear Ribonucleoprotein (A1-like) |
|---|---|
| Clone | S05-8G1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692804
|
Novus Biologicals
NBP3-19959-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671775
|
Novus Biologicals
NBP3-19959-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Heterogeneous Nuclear Ribonucleoprotein (A1-like) Monoclonal antibody specifically detects Heterogeneous Nuclear Ribonucleoprotein (A1-like) in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Heterogeneous Nuclear Ribonucleoprotein (A1-like) | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 144983 | |
| IgG | |
| Affinity purified |
| S05-8G1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| heterogeneous nuclear ribonucleoprotein A1-like 2, hnRNP A1-like 2, hnRNP core protein A1-like 2, HNRNPA1L, LOC144983, MGC102957 | |
| A synthetic peptide of human Heterogeneous Nuclear Ribonucleoprotein (A1-like) (Uniprot # Q5TBS2) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title