missing translation for 'onlineSavingsMsg'
Learn More

Hetacillin, TRC

Product Code. 30399252
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399252 1 g
30404425 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399252 Supplier Toronto Research Chemicals Supplier No. TRCH2895701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3511-16-8
Molecular Formula C19 H23 N3 O4 S
Molecular Weight (g/mol) 389.47
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[(4R)-2,2-dimethyl-5-oxo-4-phenyl-1-imidazolidinyl]-3,3-dimethyl-7-oxo-, (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-(2,2-dimethyl-5-oxo-4-phenyl-1-imidazolidinyl)-3,3-dimethyl-7-oxo-, D-(-)- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-(2,2-dimethyl-5-oxo-4-phenyl-1-imidazolidinyl)-3,3-dimethyl-7-oxo-, [2S-[2α,5α,6β(S*)]]-, (2S,5R,6R)-6-[(4R)-2,2-Dimethyl-5-oxo-4-phenyl-1-imidazolidinyl]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, BL-P 804, BRL 804, Etacillin, Hetacillin, Penplenum, Phenazacillin, Versapen, Versatrex, Vimocilin
IUPAC Name (2S,5R,6R)-6-[(4R)-2,2-dimethyl-5-oxo-4-phenylimidazolidin-1-yl]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)S[C@@H]2[C@H](N3C(=O)[C@H](NC3(C)C)c4ccccc4)C(=O)N2[C@H]1C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H23N3O4S/c1-18(2)13(17(25)26)21-15(24)12(16(21)27-18)22-14(23)11(20-19(22,3)4)10-8-6-5-7-9-10/h5-9,11-13,16,20H,1-4H3,(H,25,26)/t11-,12-,13+,16-/m1/s1
Formula Weight 389.1409
Chemical Name or Material Hetacillin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.