missing translation for 'onlineSavingsMsg'
Learn More

Hesperin, MedChemExpress

Product Code. 16439876
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439876

Brand: Medchem Express HY101371

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Hesperin is a bioactive ingredient present in Japanese horseradish (wasabi) and has been shown to be an Nrf2 activator.

Hesperin is a bioactive ingredient present in Japanese horseradish (wasabi) and has been shown to be an Nrf2 activator.

Specifications

Chemical Name or Material Hesperin
Physical Form Liquid
CAS 4430-35-7
Color Colorless
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C8H15NOS2
For Use With (Application) Cancer-programmed cell death
Solubility Information Ethanol : 100 mg/mL (487.00 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (243.50 mM; Need ultrasonic)
SMILES O=S(CCCCCCN=C=S)C
Molecular Weight (g/mol) 205.34
Formula Weight 205.34
Percent Purity 98.12%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.