missing translation for 'onlineSavingsMsg'
Learn More

Hesperidin, 95%, Thermo Scientific™

Acts as an antioxidant

£39.60 - £101.00

Chemical Identifiers

CAS 520-26-3
Molecular Formula C28H34O15
Molecular Weight (g/mol) 610.57
MDL Number MFCD00075663
InChI Key QUQPHWDTPGMPEX-QJBIFVCTSA-N
Synonym Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
PubChem CID 45358131
IUPAC Name (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494559
View Documents
Thermo Scientific Alfa Aesar
J62126.14
25g
£39.60
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15404569
View Documents
Thermo Scientific Alfa Aesar
J62126.22
100g
£101.00
100g
View alternative products
Save Up to 
»
Estimated Shipment: 11-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

Solubility
Insoluble in water. Soluble in organic solvents such as DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents. Keep away from strong light and heat.
Specifications

Chemical Identifiers

520-26-3
610.57
QUQPHWDTPGMPEX-QJBIFVCTSA-N
45358131
COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
C28H34O15
MFCD00075663
Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one

Specifications

520-26-3
MFCD00075663
Hygroscopic
Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
QUQPHWDTPGMPEX-QJBIFVCTSA-N
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
45358131
95%
C28H34O15
75140
14,4671
Insoluble in water. Soluble in organic solvents such as DMSO.
COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
610.57
610.56
Hesperidin
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-288-1

missing translation for 'rtecsNumber' : MK6650000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.