missing translation for 'onlineSavingsMsg'
Learn More

Hesperetin 3'-(2,3,4-Tri-O-acetyl-Beta-D-glucuronide Methyl Ester), TRC

Product Code. 30391210
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391210 5 mg 5mg
30394247 10 mg 10mg
30389851 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391210 Supplier Toronto Research Chemicals Supplier No. TRCH9468785MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hesperetin 3'-(2,3,4-Tri-O-acetyl-beta-D-glucuronide Methyl Ester)
Analyte or Component Names Hesperetin 3'-(2,3,4-Tri-O-acetyl-beta-D-glucuronide Methyl Ester)
Shipping Condition Room Temperature
Molecular Formula C29H30O15
Synonym (2S,3R,4S,5S,6S)-2-(5-((S)-5,7-Dihydroxy-4-oxochroman-2-yl)-2-methoxyphenoxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl Triacetate
InChI Formula InChI=1S/C29H30O15/c1-12(30)39-24-25(40-13(2)31)27(41-14(3)32)29(44-26(24)28(36)38-5)43-21-8-15(6-7-19(21)37-4)20-11-18(35)23-17(34)9-16(33)10-22(23)42-20/h6-10,20,24-27,29,33-34H,11H2,1-5H3/t20-,24-,25-,26-,27+,29+/m0/s1
Solution Type Neat
SMILES OC1=CC(O)=C(C(C[C@@H](C2=CC(O[C@H]3[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](C(OC)=O)O3)=C(OC)C=C2)O4)=O)C4=C1
IUPAC Name (2S,3R,4S,5S,6S)-2-(5-((S)-5,7-dihydroxy-4-oxochroman-2-yl)-2-methoxyphenoxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 618.54
Quantity 5 mg
Formula Weight 618.16
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.