missing translation for 'onlineSavingsMsg'
Learn More

Hesperadin, TRC

Product Code. 30400303
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400303 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400303 Supplier Toronto Research Chemicals Supplier No. TRCH2890005MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hesperadin
CAS 422513-13-1
Molecular Formula C29 H32 N4 O3 S
Quantity 5 mg
Synonym N-[2,3-Dihydro-2-oxo-3-[(3Z)-phenyl[[4-(1-piperidinylmethyl)phenyl]amino]methylene]-1H-indol-5-yl]-ethanesulfonamide, N-[(3Z)-2,3-Dihydro-2-oxo-3-[phenyl[[4-(1-piperidinylmethyl)phenyl]amino]methylene]-1H-indol-5-yl]ethanesulfonamide, Hesperadine
InChI Formula InChI=1S/C29H32N4O3S/c1-2-37(35,36)32-24-15-16-26-25(19-24)27(29(34)31-26)28(22-9-5-3-6-10-22)30-23-13-11-21(12-14-23)20-33-17-7-4-8-18-33/h3,5-6,9-16,19,30,32H,2,4,7-8,17-18,20H2,1H3,(H,31,34)/b28-27-
SMILES CCS(=O)(=O)Nc1ccc2NC(=O)\C(=C(/Nc3ccc(CN4CCCCC4)cc3)\c5ccccc5)\c2c1
IUPAC Name N-[(3Z)-2-oxo-3-[phenyl-[4-(piperidin-1-ylmethyl)anilino]methylidene]-1H-indol-5-yl]ethanesulfonamide
Molecular Weight (g/mol) 516.65
Formula Weight 516.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.