missing translation for 'onlineSavingsMsg'
Learn More

Hesperadin, MedChemExpress

Product Code. 16454958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454958 10 mg 10mg
16427697 5 mg 5mg
16421207 50 mg 50mg
16405736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16454958 Supplier Medchem Express Supplier No. HY12054
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hesperadin is an ATP competitive indolinone inhibitor of Aurora A and B. Hesperadin inhibits Aurora B with an IC50 of 250 nM. Hesperadin inhibits the growth of Trypanosoma brucei by blocking nuclear division and cytokinesis. Hesperadin also is a broad-spectrum influenza antiviral.

Hesperadin is an ATP competitive indolinone inhibitor of Aurora A and B. Hesperadin inhibits Aurora B with an IC50 of 250 nM. Hesperadin inhibits the growth of Trypanosoma brucei by blocking nuclear division and cytokinesis. Hesperadin also is a broad-spectrum influenza antiviral.

Specifications

Chemical Name or Material Hesperadin
Physical Form Solid
CAS 422513-13-1
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C29H32N4O3S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 100 mg/mL (193.55 mM)
SMILES O=C1NC2=CC=C(C=C2/C1=C(NC3=CC=C(C=C3)CN4CCCCC4)\C5=CC=CC=C5)NS(CC)(=O)=O
Molecular Weight (g/mol) 516.65
Formula Weight 516.65
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.