missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Hesperadin hydrochloride
GREENER_CHOICE

Product Code. 18095022
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095022 50 mg 50mg
18054692 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095022 Supplier Tocris Bioscience™ Supplier No. 3988/50
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent Aurora kinase B inhibitor

ATP-competitive inhibitor of Aurora B kinase (IC50 = 250 nM). Prevents chromosome alignment and segregation; also induces polyploidy and prevents histone H3-Ser10 phosphorylation. Overrides the spindle assembly checkpoint and induces mitotic exit in monastrol - and taxol -treated HeLa cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422513-13-1
Molecular Formula C29H32N4O3S

Specifications

Quantity 50 mg
Formula Weight 553.12
Purity >99%
Molecular Weight (g/mol) 516.66
Chemical Name or Material Hesperadin hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.