missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES6 Antibody (OTI1D6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72008F
This item is not returnable.
View return policy
Description
HES6 Monoclonal antibody specifically detects HES6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| HES6 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1D6 | |
| Western Blot | |
| BHLHB41, bHLHb41Class B basic helix-loop-helix protein 41, bHLHc23, C-HAIRY1, Hairy and enhancer of split 6, hairy and enhancer of split 6 (Drosophila), HES-6, transcription cofactor HES-6 | |
| Full length human recombinant protein of human HES6 (NP_061115) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 55502 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction