missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541167
This item is not returnable.
View return policy
Description
Peptide sequence: MAADTPGKPS ASPMAGAPAS ASRTPDKPRS AAEHRKSSKP VMEKRRRARI Sequence homology: Cow: 86%; Dog: 93%; Human: 100%; Pig: 85%; Rat: 77%.
HES4, or Hes Family BHLH Transcription Factor 4, is a gene located on chromosome 1p36.31, encoding a basic helix-loop-helix (bHLH) transcription factor that plays a role in the Notch signaling pathway. This pathway is integral to cell differentiation, proliferation, and apoptotic processes, especially during embryonic development and tissue homeostasis. HES4 acts as a transcriptional repressor, regulating the expression of genes responsible for maintaining stem cell populations and inhibiting premature differentiation. While its exact biological roles are still being elucidated, studies suggest HES4 influences neurogenesis and myogenesis by modulating the timing of differentiation events within progenitor cells. The dysregulation of HES4 can impact normal developmental processes, potentially contributing to pathologies such as cancer and developmental disorders due to its control over cellular growth and differentiation pathways. Research into HES4 continues to advance understanding of its functions within the context of Notch signaling and its implications for disease.
Specifications
| HES4 | |
| Polyclonal | |
| Unconjugated | |
| HES4 | |
| bHLH factor Hes4; bHLHb42; Class B basic helix-loop-helix protein 42; hairy and enhancer of split 4; hes family bHLH transcription factor 4; HES4; hHES4; transcription factor HES-4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 57801 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HCC6 | |
| HES4 | |
| Synthetic peptide directed towards the N-terminal of human HES4 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction