missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HES4 Monoclonal Antibody (455237)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561328

Product Code. 30535280

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

HES4, or Hes Family BHLH Transcription Factor 4, is a gene located on chromosome 1p36.31, encoding a basic helix-loop-helix (bHLH) transcription factor that plays a role in the Notch signaling pathway. This pathway is integral to cell differentiation, proliferation, and apoptotic processes, especially during embryonic development and tissue homeostasis. HES4 acts as a transcriptional repressor, regulating the expression of genes responsible for maintaining stem cell populations and inhibiting premature differentiation. While its exact biological roles are still being elucidated, studies suggest HES4 influences neurogenesis and myogenesis by modulating the timing of differentiation events within progenitor cells. The dysregulation of HES4 can impact normal developmental processes, potentially contributing to pathologies such as cancer and developmental disorders due to its control over cellular growth and differentiation pathways. Research into HES4 continues to advance understanding of its functions within the context of Notch signaling and its implications for disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HES4
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
Q9HCC6
HES4
E. coli-derived recombinant human HES-4, Val94-Arg221.
100 μg
Primary
Human
Antibody
IgG2a
Western Blot
455237
Unconjugated
HES4
bHLH factor Hes4; bHLHb42; Class B basic helix-loop-helix protein 42; hairy and enhancer of split 4; hes family bHLH transcription factor 4; HES4; hHES4; transcription factor HES-4
Mouse
Protein A/G
RUO
57801
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.