missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567801
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Transcriptional repressor of genes that require a bHLH protein for their transcription. Target protein's helix-loop-helix (HLH) protein; may function as repressor in neurons.
Specifications
| HES3 | |
| Polyclonal | |
| Unconjugated | |
| HES3 | |
| BHLHB43; Class B basic helix-loop-helix protein 43; hairy and enhancer of split 3; hairy and enhancer of split 3 (Drosophila); hes family bHLH transcription factor 3; HES3; Hes-3; Transcription factor HES-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15207, 390992, 64628 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q04667, Q5TGS1, Q61657 | |
| HES3 | |
| Synthesized peptide derived from human HES3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction