missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588220
This item is not returnable.
View return policy
Description
Immunogen sequence: NAMTYPGQPH PALQAPPPPP PGPGGPQHAP FAPPPPLVPI PGGAAPPPGG APCKLGSQAG EAAKVFGGFQ VVPAPDGQFA FLIPNGAFAH SGPVIPVYTS NSGTSVGPNA VSPSSGPSLT ADSMWRPWRN; Positive Samples: Rat testis; Cellular Location: Nucleus.
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.
Specifications
| HES1 | |
| Polyclonal | |
| Unconjugated | |
| HES1 | |
| BHLHB39; class B basic helix-loop-helix protein 39; FLJ20408; Hairy and enhancer of split 1; hairy and enhancer of split 1 (Drosophila); hairy and enhancer of split 1,; hairy homolog; Hairy-like; Hairy-like protein; hes family bHLH transcription factor 1; HES1; Hes-1; hHL; HL; HRY; RHL; transcription factor HES-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29577 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.02 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q04666 | |
| HES1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 151-280 of human HES1 (NP_0055151). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction