missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102850
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HES1.
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.
Specifications
| HES1 | |
| Polyclonal | |
| Unconjugated | |
| HES1 | |
| BHLHB39; class B basic helix-loop-helix protein 39; FLJ20408; Hairy and enhancer of split 1; hairy and enhancer of split 1 (Drosophila); hairy and enhancer of split 1,; hairy homolog; Hairy-like; Hairy-like protein; hes family bHLH transcription factor 1; HES1; Hes-1; hHL; HL; HRY; RHL; transcription factor HES-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15205, 29577, 3280 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35428, Q04666, Q14469 | |
| HES1 | |
| A synthesized peptide derived from human HES1(Accession Q14469), corresponding to amino acid residues D63-G113. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction