missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES-1 Antibody (OTI4H1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70937MFV450
This item is not returnable.
View return policy
Description
HES-1 Monoclonal antibody specifically detects HES-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| HES-1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI4H1 | |
| Flow Cytometry | |
| BHLHB39, bHLHb39hairy homolog (Drosophila), Class B basic helix-loop-helix protein 39, FLJ20408, Hairy and enhancer of split 1, hairy and enhancer of split 1, (Drosophila), Hairy homolog, Hairy-like protein, Hes1, HES-1, hHL, HL, HRYHHL, transcription factor HES-1 | |
| Recombinant protein expressed in E.coli corresponding to amino acids 1-280 of human HES1 | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3280 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction