missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HERV Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522819
This item is not returnable.
View return policy
Description
HERV Polyclonal Antibody for Western Blot, IHC (P), IHC (F), Neu
HERVs are a class of multiple sclerosis associated retroviruses. Reacts with a replication competent Human Endogenous Retrovirus (HERVW) isolated from placenta. The synthetic peptide sequence used as immunogen is taken from the virus envelope polyprotein sequence. Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. The HERVW endogenous envelope protein has retained its original fusogenic properties and participates in trophoblast fusion during placenta morphogenesis.
Specifications
| HERV | |
| Polyclonal | |
| Unconjugated | |
| ERVW-1 | |
| endogenous retroviral family W, env(C7), member 1; endogenous retrovirus group W member 1; endogenous retrovirus group W, member 1; ENV; envelope glycoprotein; envelope polyprotein gPr73; envelope protein; Enverin; ENVW; Env-W; ERVW-1; ERVWE1; gp24; gp50; HERV7Q; HERV-7q; HERV-7q envelope protein; HERV-tryptophan envelope protein; HERVW; HERV-W Env glycoprotein; HERV-W envelope protein; HERV-W_7q21.2 provirus ancestral Env polyprotein; HERVWENV; HERV-W-ENV; human endogenous retrovirus W envC7-1 envelope protein; SU; Surface protein; Syncytin; syncytin A; syncytin-1; TM; Transmembrane protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30816 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9UQF0 | |
| ERVW-1 | |
| Synthetic peptide corresponding to residues 42-50 of the HERV protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction