Learn More
Invitrogen™ HERPUD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109809
Description
Immunogen sequence: MESETEPEPV TLLVKSPNQR HRDLELSGDR GWSVGHLKAH LSRVYPERPR PEDQRLIYSG KLLLDHQCLR DLLPKQEKRH VLHLVCNVKS PSKMPEINAK VAESTEEPAG SNRGQYPEDS SSDGLRQREV LRNLSSPGWE NISRPEAAQQ AFQGLGPGFS GYTPYGWLQL SWFQQIYARQ YYMQYLAATA ASGAFVPPPS AQEIPVVSAP APAPIHNQFP AENQPANQNA APQVVVNPGA.
The accumulation of unfolded proteins in the endoplasmic reticulum (ER) triggers the ER stress response. This response includes the inhibition of translation to prevent further accumulation of unfolded proteins, the increased expression of proteins involved in polypeptide folding, known as the unfolded protein response (UPR), and the destruction of misfolded proteins by the ER-associated protein degradation (ERAD) system. This gene may play a role in both UPR and ERAD. Its expression is induced by UPR and it has an ER stress response element in its promoter region while the encoded protein has an N-terminal ubiquitin-like domain which may interact with the ERAD system. This protein has been shown to interact with presenilin proteins and to increase the level of amyloid-beta protein following its overexpression. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined.
Specifications
| HERPUD1 | |
| Polyclonal | |
| Unconjugated | |
| HERPUD1 | |
| Herp; HERPUD1; homocysteine inducible ER protein with ubiquitin like domain 1; homocysteine-inducible endoplasmic reticulum stress-inducible ubiquitin-like domain member 1 protein; homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1; homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domain member 1 protein; KIAA0025; Methyl methanesulfonate (MMF)-inducible fragment protein 1; Mif1; Mifl; MMS-inducible; Sup | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64209, 85430, 9709 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15011, Q9JJK5 | |
| HERPUD1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human HERPUD1 (NP_055500.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.