missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Herpes Simplex Virus VP16 Polyclonal antibody specifically detects Herpes Simplex Virus VP16 in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Herpes Simplex Virus VP16 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to amino acids 474-487 of the herpes simplex virus Herpes Simplex Virus VP16 protein, conjugated to KLH via an N-terminal added cysteine. |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?