missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes simplex virus gB Mouse anti-Herpes simplex virus, Clone: P1123, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13568.500ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against herpes simplex virus (HSV) gB.
- High specificity
- High reactivity
Specifications
| Herpes simplex virus gB | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence Western Blot The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 500 μg | |
| Primary | |
| Herpes Simplex virus (HSV) | |
| Liquid |
| Immunofluorescence, Western Blot | |
| P1123 | |
| Mouse monoclonal antibody raised against herpes simplex virus (HSV) gB. | |
| In PBS, pH 7.4. | |
| Protein G purification | |
| RUO | |
| This antibody reacts with gB of herpes simplex virus. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction