missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 2 Antibody (T303), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-478AF594
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 2 Monoclonal antibody specifically detects Herpes Simplex Virus 2 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications
| Herpes Simplex Virus 2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| The antibody T303 recognizes gG antigen of HSV2 (Herpes Simplex Virus type 2), a dsDNA virus, member of Herpesviridae family. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| T303 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Herpes Simplex Virus Type 2 | |
| Exctract of Herpes Simplex Virus 2 infected VERO (green monkey kidney) cells. | |
| Primary | |
| Virus | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto