missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 1 (HSV1) Mouse anti-Virus, Clone: HSV1/1934, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Herpes Simplex Virus 1 (HSV1) Monoclonal specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Herpes Simplex Virus 1 (HSV1) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HSV1/1934 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?