missing translation for 'onlineSavingsMsg'
Learn More

Herpes Simplex Virus 1 Antibody (HSVI/2045), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18373672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373672 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373672 Supplier Novus Biologicals Supplier No. NBP308639JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Herpes Simplex Virus 1 (HSV1) Monoclonal antibody specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Herpes Simplex Virus 1 (HSV1)
Applications Western Blot
Classification Monoclonal
Clone HSVI/2045
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2
Host Species Mouse
Immunogen Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.