missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 1 Antibody (10A3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08625CL1
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Herpes Simplex Virus 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1 | |
| Detergent-solubilized Herpes Simplex Virus 1 infected cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| 10A3 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction