missing translation for 'onlineSavingsMsg'
Learn More

HERC5 Antibody, Novus Biologicals™

Product Code. 18342309 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342309 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342309 Supplier Novus Biologicals Supplier No. H00051191A01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

HERC5 Polyclonal antibody specifically detects HERC5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC5
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation 50% Glycerol
Gene Accession No. NP_057407
Gene Alias CEB1E3 ISG15--protein ligase HERC5, CEBP1, cyclin-E binding protein 1, Cyclin-E-binding protein 1, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 5, hect domain and RLD 5, HECT E3 ubiquitin ligase, probable E3 ubiquitin-protein ligase HERC5
Host Species Mouse
Immunogen HERC5 (NP_057407, 915 a.a. - 1024 a.a.) partial recombinant protein with GST tag. YDWKTFEKNARYEPGYNSSHPTIVMFWKAFHKLTLEEKKKFLVFLTGTDRLQMKDLNNMKITFCCPESWNERDPIRALTCFSVLFLPKYSTMETVEEALQEAINNNRGFG
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51191
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.