missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98017-50ul
This item is not returnable.
View return policy
Description
HERC2 Polyclonal antibody specifically detects HERC2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HERC2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| D15F37S1, DKFZp547P028, E3 ubiquitin-protein ligase HERC2, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 2, hect domain and RLD 2, jdf2, KIAA0393, p528, probable E3 ubiquitin-protein ligase HERC2, SHEP1 | |
| Produced in rabbits immunized with E. coli-derived Human HERC2 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8924 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction