missing translation for 'onlineSavingsMsg'
Learn More

HERC2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18424333 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424333 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424333 Supplier Novus Biologicals Supplier No. NBP105970AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HERC2 Polyclonal antibody specifically detects HERC2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias D15F37S1, DKFZp547P028, E3 ubiquitin-protein ligase HERC2, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 2, hect domain and RLD 2, jdf2, KIAA0393, p528, probable E3 ubiquitin-protein ligase HERC2, SHEP1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 4784 and 4834 of human hect domain and RLD 2 using the numbering given in entry NP_004658.3 (GeneID 8924).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8924
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.