missing translation for 'onlineSavingsMsg'
Learn More

HERC1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18432553 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432553 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432553 Supplier Novus Biologicals Supplier No. NBP105969JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HERC1 Polyclonal antibody specifically detects HERC1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2, guanine nucleotide exchange factor p532, hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1(CHC1)-like domain (RLD) 1, hect domain and RCC1-like domain 1, HECT domain and RCC1-like domain-containing protein 1, p532probable E3 ubiquitin-protein ligase HERC1, p619EC 6.3.2.-
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2400 and 2450 of human hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 1 using the numbering given in entry NP_003913.3 (GeneID 8
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8925
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.