missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HER2 Trastuzumab297 anti-Human, BUV395, Clone: Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb), BD OptiBuild™
Shop All BD Biosciences ProductsDescription
HER2 Trastuzumab297 Monoclonal antibody specifically detects HER2 Trastuzumab297 in Human samples. It is validated for Flow cytometry
Specifications
Specifications
| Antigen | HER2 Trastuzumab297 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb) |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Ultraviolet 395 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CD340; ERBB2; HER-2; HER-2/neu; HER2; NEU; proto-oncogene c-ErbB-2 |
| Immunogen | A431 Cell Line Expressing EGFR |
| Purification Method | Affinity Chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?