missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HER2 PROTAC™ CH7C4

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
HER2 PROTAC™ CH7C4 is a potent and selective HER2 (also called ErbB-2) targeting PROTAC™ (DC50 = 69 nM; Dmax = 96%). Inhibits BT-474 cell proliferation (IC50 = 47 nM) and xenograft growth. Comprises a Tucatinib (Cat. No. 7640) analog, joined by a linker to a CRBN E3 ligase ligand. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 5-((7-(4-(3-((4-((4-([1,2,4]Triazolo[1,5-a]pyridin-7-yloxy)-3-methylphenyl)amino)-6-methoxyquinazolin-7-yl)oxy)propyl)piperazin-1-yl)heptyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C49H55N11O7 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction