missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Her2/Neu Mouse anti-Human, BV750, Clone: NEU 24.7, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747064
This item is not returnable.
View return policy
Description
Her2/Neu Monoclonal antibody specifically detects Her2/Neu in Human samples. It is validated for Flow Cytometry
Specifications
| Her2/Neu | |
| Monoclonal | |
| Brilliant Violet 750 | |
| HER2; NEU; ERBB2; p185HER2; erb-b2 receptor tyrosine kinase 2; CD340 | |
| SKBR3 human breast carcinoma cells | |
| RUO | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| NEU 24.7 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction