missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Her2/Neu Mouse anti-Human, BV480, Clone: NEU 24.7, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 746442
This item is not returnable.
View return policy
Description
Her2/Neu Monoclonal antibody specifically detects Her2/Neu in Human samples. It is validated for Flow Cytometry.
Specifications
| Her2/Neu | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| NEU 24.7 | |
| Brilliant Violet 480 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction