Learn More
HER-2/c-erbB-2/neu Ab-17 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (For cytoplasmic), Epredia™ HER-2/c-erbB-2/neu Ab-17, Mouse Monoclonal Antibody provides accurate, reproducible results..
Brand: Epredia MS-730-P1
Description
c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the proteinHost Species: Mouse
Clone: e2-4001 + 3B5
Isotype: IgG1 + IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Cytoplasmic
Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human HER-2 protein (3B5)
Molecular Weight: 185kDa
Positive Control: LS174T cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (For cytoplasmic)
Specifications
| HER-2/c-erbB-2/neu Ab-17 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| EGFR Family | |
| Human, Rat | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| e2-4001 + 3B5 | |
| Unconjugated | |
| Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5) | |
| ASR, IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.