missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptyl chloroformate, TRC
Specifications
Specifications
| Chemical Name or Material | Heptyl chloroformate |
| CAS | 33758-34-8 |
| Molecular Formula | C8 H15 Cl O2 |
| Quantity | 100 mg |
| Synonym | n-Heptyl chloroformate, Heptyl chlorocarbonate, 1-Heptyl chloroformate |
| InChI Formula | InChI=1S/C8H15ClO2/c1-2-3-4-5-6-7-11-8(9)10/h2-7H2,1H3 |
| SMILES | CCCCCCCOC(=O)Cl |
| IUPAC Name | heptyl carbonochloridate |
| Molecular Weight (g/mol) | 178.656 |
| Formula Weight | 178.076 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?