missing translation for 'onlineSavingsMsg'
Learn More

Heptyl beta-D-glucopyranoside, 99%

Product Code. 15417989
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417989 500 mg 500mg
15407989 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417989 Supplier Thermo Scientific Alfa Aesar Supplier No. J66576.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78617-12-6 | C13H26O6 | 278.345 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78617-12-6
Molecular Formula C13H26O6
Molecular Weight (g/mol) 278.345
MDL Number MFCD00063299
InChI Key NIDYWHLDTIVRJT-UJPOAAIJSA-N
Synonym heptyl-beta-d-glucopyranoside, heptyl beta-d-glucopyranoside, 2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, b7g, n-heptyl beta-d-glucopyranoside, n-heptyl-i 2-d-glucopyranoside, heptyl-?-d-glucopyranoside, heptyl b-d-glucopyranoside, heptyl-b-d-glucopyranoside, d06ibv
PubChem CID 448173
IUPAC Name (2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 500 mg
Beilstein 8806
Formula Weight 278.34
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Heptyl beta-D-glucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.