missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Heptanoic acid (Heptylic acid) |
| CAS | 111-14-8 |
| Molecular Formula | C7 H14 O2 |
| Quantity | 250 g |
| Synonym | Heptanoic Acid, 1-Hexanecarboxylic acid, Enanthic acid, Enanthylic acid, Enantic acid, Heptoic acid, Heptylic acid, NSC 2192, Oenanthic acid, Oenanthylic acid, n-Heptanoic acid, n-Heptoic acid, n-Heptylic acid |
| InChI Formula | InChI=1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9) |
| SMILES | CCCCCCC(=O)O |
| IUPAC Name | heptanoic acid |
| Molecular Weight (g/mol) | 130.18 |
| Formula Weight | 130.0994 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?