missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic Acid, Mikromol™

Product Code. 30052277
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052277 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052277 Supplier Mikromol Supplier No. MM1119.010100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Heptanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heptanoic acid (Heptylic acid)
CAS 111-14-8
Analyte or Component Names Heptanoic acid (Heptylic acid)
Shipping Condition Room Temperature
Molecular Formula C7 H14 O2
Certifications/Compliance ISO 9001
Synonym Heptanoic Acid, 1-Hexanecarboxylic acid, Enanthic acid, Enanthylic acid, Enantic acid, Heptoic acid, Heptylic acid, NSC 2192, Oenanthic acid, Oenanthylic acid, n-Heptanoic acid, n-Heptoic acid, n-Heptylic acid
InChI Formula InChI=1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)
SMILES CCCCCCC(=O)O
IUPAC Name heptanoic acid
Molecular Weight (g/mol) 130.18
Quantity 100 mg
Formula Weight 130.0994 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.