missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic acid, 98+%

Product Code. 11403288
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403288 500 mL 500mL
11413288 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403288 Supplier Thermo Scientific Alfa Aesar Supplier No. A17704.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-14-8 | C7H14O2 | 130.187 g/mol

Heptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.

Solubility
Miscible with acetone, ethanol, diethyl ether, dimethylformamide, dimethyl sulfoxide and nitric acid. Slightly soluble in carbon tetrachloride and water.

Notes
Incompatible with strong oxidizing agents, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-14-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00004426
InChI Key MNWFXJYAOYHMED-UHFFFAOYSA-N
Synonym enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
PubChem CID 8094
ChEBI CHEBI:45571
IUPAC Name heptanoic acid
SMILES CCCCCCC(=O)O

Specifications

Melting Point -8°C
Density 0.917
Boiling Point 222°C to 223°C
Flash Point 114°C (237°F)
Linear Formula CH3(CH2)5CO2H
Refractive Index 1.423
Quantity 500 mL
UN Number UN3265
Beilstein 1744723
Merck Index 14,4660
Solubility Information Miscible with acetone,ethanol,diethyl ether,dimethylformamide,dimethyl sulfoxide and nitric acid. Slightly soluble in carbon tetrachloride and water.
Formula Weight 130.19
Percent Purity ≥98%
Chemical Name or Material Heptanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.