missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic acid, 98%

Product Code. 10092000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092000 1 L Glass bottle 1 L
10783351 250 mL Glass bottle 250mL
13468398 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092000 Supplier Thermo Scientific Acros Supplier No. 164170010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-14-8 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-14-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00004426
InChI Key MNWFXJYAOYHMED-UHFFFAOYSA-N
Synonym enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
PubChem CID 8094
ChEBI CHEBI:45571
IUPAC Name heptanoic acid
SMILES CCCCCCC(=O)O

Specifications

Melting Point -10.5°C
Color White
Density 0.9100g/mL
Boiling Point 223.0°C to 223.4°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)5CO2H
Refractive Index 1.4210 to 1.4240
Quantity 1 L
Beilstein 02,338
Merck Index 15,4695
Specific Gravity 0.91
Solubility Information Solubility in water: 0.24g/100mL (15°C). Other solubilities: soluble in alcohol, ether, dmf, chloroform, petrol, soluble in most organic solvents
Viscosity 3.40 mPa.s (30°C)
Formula Weight 130.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Heptanoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Heptanoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Heptanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.