missing translation for 'onlineSavingsMsg'
Learn More

Heptane, Honeywell Riedel-de Haën™

Product Code. 15915430
Change view
Click to view available options
Quantity:
200 L
Packaging:
Returnable Stainless Steel Drum
Grade:
HPLC
Unit Size:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Grade unitSize
15915430 200 L Returnable Stainless Steel Drum HPLC 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15915430 Supplier Honeywell-Riedel-de Haen Supplier No. 04882200L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

For preparative HPLC, ≥95.0% (GC)

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -90.5°C
CAS 142-82-5
Color Colorless
Quantity 200 L
Packaging Returnable Stainless Steel Drum
Molecular Formula C7H16
MDL Number MFCD00009544
Synonym n-Heptane, Heptanes
Boiling Point 98.2°C to 98.4°C (1.000 hPa)
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
Grade HPLC
Chemical Name or Material Heptane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.