missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC), Honeywell Riedel-de Haën™
Puriss. p.a., Reag. Ph. Eur.,≥99% n-heptane basis (GC)
£356.15 - £728.36
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15613760
|
Honeywell-Riedel-de Haen
32287-6X1L |
6 x 1 L | Not Available |
£356.15
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15683750
|
Honeywell-Riedel-de Haen
32287-4X2.5L |
4 x 2.5 L | Glass Bottle |
£617.45
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15693750
|
Honeywell-Riedel-de Haen
32287-4X5L |
4 x 5 L | Aluminum Bottle | ||||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
-91°C | |
Colorless liquid | |
98.2°C to 98.4°C | |
−4°C (24.8°F) | |
n20/D 1.3870 to 1.3880 | |
C7H16 | |
CH3(CH2)5CH3 | |
40mmHg (20°C), 83mmHg (37.7°C) | |
1730763 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2g/mol | |
puriss. p.a. |
142-82-5 | |
0.684g/mL at 25°C | |
0.07 | |
433°F | |
0 to 0.001% | |
Free acid (as CH3COOH): ≤0.0005% | |
MFCD00009544 | |
UN1206 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
≥99% n to heptane basis (GC) | |
Heptane, Reag. Ph. Eur. |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
H225-H304-H315-H336-H410
Danger
missing translation for 'dotInformation' : Class 3 - PG 2 - Heptanes
missing translation for 'einecsNumber' : 205-563-8
missing translation for 'rtecsNumber' : MI7700000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- n-Heptane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.