missing translation for 'onlineSavingsMsg'
Learn More

Solstice Heptane Fraction

Product Code. 15985440
Change view
Click to view available options
Quantity:
7 L
18 L
45 L
Unit Size:
18 L
45 L
7 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15985440 45 L 45 L
15655705 7 L 7 L
15975440 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15985440 Supplier Solstice Supplier No. 3200445L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Heptane Fraction, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC).

Supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:

  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-Heptane, Heptanes
IUPAC Name heptane
SMILES CCCCCCC

Specifications

pH Not applicable
Melting Point -90.5°C
Color Colorless
Density 0.69 g/cm3 (15°C)
Quantity 45 L
Flash Point 6.7 %(V)
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Vapor Pressure 60.9 hPa (25°C)
Viscosity No data available
UN Number UN1206
Beilstein 1730763
Boiling Point 98.2°C to 98.4°C (1.000 hPa)
Percent Purity ≥99.00%
Grade Reagent
Chemical Name or Material Heptane Fraction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.