missing translation for 'onlineSavingsMsg'
Learn More

Heptanal and Nonanal Mixture 671 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30325051
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325051 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325051 Supplier Dr. Ehrenstorfer Supplier No. DREA50000671ME
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 1-Heptanal∣Pelargonaldehyde
CAS 111-71-7, 124-19-6
Analyte or Component Names 1-Heptanal∣Pelargonaldehyde
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1-Heptanal 100 μg/mL∣Pelargonaldehyde 100 μg/mL
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.