missing translation for 'onlineSavingsMsg'
Learn More

Heptan-1-ol, 98+%, Fisher Chemical

Product Code. 10745331 Shop All Fisher Chemical Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
missing translation for 'unitSize':
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
10745331 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10745331 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' H/0250/05

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 111-70-6
Melting Point -35°C
Boiling Point 176°C
Packaging Amber glass bottle
Molecular Formula C7H16O
MDL Number MFCD00002986
Quantity 100 mL
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
SMILES CCCCCCCO
IUPAC Name heptan-1-ol
Molecular Weight (g/mol) 116.20
PubChem CID 8129
Formula Weight 116.2g/mol
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • 1-Heptanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.