missing translation for 'onlineSavingsMsg'
Learn More

Heptakis-6-bromo-6-deoxy-beta-cyclodextrin, Thermo Scientific™

Product Code. 10105370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10105370 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10105370 Supplier Thermo Scientific Acros Supplier No. 299005000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 53784-83-1
Melting Point 217°C to 221°C
Color White
Packaging Glass bottle
Molecular Formula C42H63Br7O28
MDL Number MFCD00803289
Quantity 500 mg
Synonym heptakis 6-bromo-6-deoxy-beta-cyclodextrin
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf,dmso,pyridine
InChI Key XNNRROJOWZLYCK-UHFFFAOYNA-N
SMILES OC1C(O)C2OC3OC(CBr)C(OC4OC(CBr)C(OC5OC(CBr)C(OC6OC(CBr)C(OC7OC(CBr)C(OC8OC(CBr)C(OC1OC2CBr)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
IUPAC Name 5,10,15,20,25,30,35-heptakis(bromomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular Weight (g/mol) 1575.27
PubChem CID 11137055
Formula Weight 1575.28
Physical Form Powder
Chemical Name or Material Heptakis-6-bromo-6-deoxy-β-cyclodextrin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.