missing translation for 'onlineSavingsMsg'
Learn More

Heptahelicene, TRC

Product Code. 30398983
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398983 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398983 Supplier Toronto Research Chemicals Supplier No. TRCH2966505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heptahelicene
CAS 16914-68-4
Molecular Formula C30 H18
Quantity 5 mg
Synonym Dinaphtho[2,1-c:1',2'-g]phenanthrene
InChI Formula InChI=1S/C30H18/c1-3-7-25-19(5-1)9-11-21-13-15-23-17-18-24-16-14-22-12-10-20-6-2-4-8-26(20)28(22)30(24)29(23)27(21)25/h1-18H
SMILES c1ccc2c(c1)ccc3ccc4ccc5ccc6ccc7ccccc7c6c5c4c23
IUPAC Name heptacyclo[16.12.0.02,15.03,12.04,9.021,30.024,29]triaconta-1,3(12),4,6,8,10,13,15,17,19,21(30),22,24,26,28-pentadecaene
Molecular Weight (g/mol) 378.464
Formula Weight 378.141
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.