missing translation for 'onlineSavingsMsg'
Learn More

(Heptafluorobutyro)phenone, 99%

Product Code. 11303746
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11303746

Brand: Thermo Scientific Alfa Aesar L16777.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 559-91-1 | C10H5F7O | 274.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 559-91-1
Molecular Formula C10H5F7O
Molecular Weight (g/mol) 274.138
MDL Number MFCD00077552
InChI Key UKYZDGAVBNKBPQ-UHFFFAOYSA-N
Synonym heptafluoropropyl phenyl ketone, heptafluorobutyrophenone, phenyl heptafluoropropyl ketone, pubchem20274, heptafluorobutyro phenone, 1-heptafluorobutyryl benzene, heptafluoropropylphenylketone, 2,2,3,3,4,4,4-heptafluoro-1-phenyl-butan-1-one
PubChem CID 238424
IUPAC Name 2,2,3,3,4,4,4-heptafluoro-1-phenylbutan-1-one
SMILES C1=CC=C(C=C1)C(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.473
Boiling Point 170°C to 172°C
Refractive Index 1.414
Quantity 5 g
Beilstein 2056234
Formula Weight 274.14
Percent Purity 99%
Chemical Name or Material (Heptafluorobutyro)phenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.